Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDX14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117268.3 | NP_567384.1 | 3 - 4 | 428 | 72 | |
GenBank mRNA | AJ748742.1 | - | 3 - 4 | 412 | 72 | |
AY052271.1 | - | 3 - 4 | 421 | 72 | ||
BT001146.1 | - | 3 - 4 | 398 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |