Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341318.1 | NP_001329724.1 | 22 - 23 | 3535 | 64 | |
NM_118020.5 | NP_193637.2 | 21 - 22 | 3714 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |