Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCED4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118036.3 | NP_193652.1 | 1 | 2055 | 132 | |
GenBank mRNA | AK227175.1 | - | 1 | 1863 | 132 | |
AY056789.1 | - | 1 | 1888 | 132 | ||
AY136353.1 | - | 1 | 1886 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |