Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343647.1 | NP_001330416.1 | 4 - 5 | 782 | 58 | |
NM_122031.4 | NP_197524.1 | 5 - 6 | 793 | 58 | ||
GenBank mRNA | D30807.1 | - | 5 - 6 | 517 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |