Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161252.2 | NP_001154724.1 | 8 - 9 | 1101 | 128 | |
NM_001343670.1 | NP_001318611.1 | 8 - 9 | 1080 | 128 | ||
NM_001343671.1 | NP_001318612.1 | 9 - 10 | 1533 | 128 | ||
GenBank mRNA | HQ427882.1 | - | 8 - 9 | 1083 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |