Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036838.2 | NP_001031915.1 | 4 - 5 | 664 | 85 | |
NM_001343687.1 | NP_001332603.1 | 4 - 5 | 464 | 85 | ||
NM_122090.4 | NP_197583.1 | 3 - 4 | 1063 | 85 | ||
GenBank mRNA | AK316826.1 | - | 3 - 4 | 470 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |