Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122101.2 | NP_568405.1 | 4 - 5 | 942 | 78 | |
GenBank mRNA | L23985.1 | - | 4 - 5 | 902 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |