Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122127.3 | NP_568412.1 | 2 - 3 | 659 | 75 | |
GenBank mRNA | AK118386.1 | - | 2 - 3 | 532 | 75 | |
BT026479.1 | - | 2 - 3 | 457 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |