Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QQT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125789.1 | NP_001119261.1 | 2 - 3 | 188 | 113 | |
NM_001343725.1 | NP_001332350.1 | 1 - 2 | 451 | 113 | ||
NM_122142.4 | NP_197629.2 | 2 - 3 | 258 | 113 | ||
GenBank mRNA | AK221541.1 | - | 2 - 3 | 157 | 113 | |
AX765846.1 | - | 1 - 2 | 86 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |