Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HUA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343791.1 | NP_001330849.1 | 5 - 6 | 2974 | 73 | |
NM_001343792.1 | NP_001330848.1 | 5 - 6 | 2882 | 73 | ||
NM_122221.3 | NP_197706.1 | 5 - 6 | 2882 | 73 | ||
GenBank mRNA | AB493755.1 | - | 5 - 6 | 2544 | 73 | |
AF116556.1 | - | 5 - 6 | 2882 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |