Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HUA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343791.1 | NP_001330849.1 | 7 - 8 | 3275 | 139 | |
NM_001343792.1 | NP_001330848.1 | 7 - 8 | 3183 | 139 | ||
NM_122221.3 | NP_197706.1 | 7 - 8 | 3183 | 139 | ||
GenBank mRNA | AB493755.1 | - | 7 - 8 | 2845 | 139 | |
AF116556.1 | - | 7 - 8 | 3183 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |