Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDPK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122264.5 | NP_197748.1 | 5 - 6 | 1524 | 113 | |
GenBank mRNA | AK229078.1 | - | 5 - 6 | 1397 | 113 | |
BT026382.1 | - | 5 - 6 | 1338 | 113 | ||
U20388.1 | - | 5 - 6 | 1375 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |