Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWEET12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122271.3 | NP_197755.1 | 3 - 4 | 503 | 70 | |
GenBank mRNA | AF095641.1 | - | 3 - 4 | 408 | 70 | |
AY057575.1 | - | 3 - 4 | 433 | 70 | ||
AY059108.1 | - | 3 - 4 | 304 | 70 | ||
AY116672.1 | - | 3 - 4 | 430 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |