Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122304.1 | NP_197787.1 | 3 - 4 | 312 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |