Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122491.3 | NP_197962.1 | 1 - 2 | 394 | 78 | |
GenBank mRNA | AF047719.1 | - | 1 - 2 | 168 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |