Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122513.4 | NP_197984.2 | 3 - 4 | 327 | 75 | |
GenBank mRNA | AY063925.1 | - | 3 - 4 | 327 | 75 | |
AY243510.1 | - | 2 - 3 | 257 | 75 | ||
BT006062.1 | - | 2 - 3 | 252 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |