Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK 11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122557.3 | NP_568486.1 | 8 - 9 | 1038 | 74 | |
GenBank mRNA | AF428327.1 | - | 8 - 9 | 1038 | 74 | |
AY046024.1 | - | 8 - 9 | 1038 | 74 | ||
AY142595.1 | - | 7 - 8 | 795 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |