Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBP60G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203479.1 | NP_001190408.1 | 3 - 4 | 863 | 70 | |
NM_001343987.1 | NP_001331616.1 | 3 - 4 | 863 | 70 | ||
NM_122574.4 | NP_198044.2 | 3 - 4 | 866 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |