Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203480.1 | NP_001190409.1 | 19 - 20 | 2776 | 67 | |
NM_122585.5 | NP_198055.3 | 20 - 21 | 2856 | 67 | ||
GenBank mRNA | BT004037.1 | - | 20 - 21 | 2687 | 67 | |
BT005165.1 | - | 20 - 21 | 2554 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |