Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AST91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332586.1 | NP_001320230.1 | 7 - 8 | 2142 | 118 | |
NM_001332587.1 | NP_001320231.1 | 8 - 9 | 2057 | 118 | ||
NM_001332588.1 | NP_001320232.1 | 8 - 9 | 2012 | 118 | ||
NM_102157.4 | NP_173722.1 | 9 - 10 | 1941 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |