Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332600.1 | NP_001320962.1 | 1 - 2 | 227 | 75 | |
NM_102191.6 | NP_564194.1 | 1 - 2 | 186 | 75 | ||
GenBank mRNA | AB493475.1 | - | 1 - 2 | 101 | 75 | |
AF195047.1 | - | 1 - 2 | 128 | 75 | ||
BT026410.1 | - | 1 - 2 | 101 | 75 | ||
DQ446281.1 | - | 1 - 2 | 101 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |