Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH2B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332622.1 | NP_001321290.1 | 3 - 4 | 425 | 110 | |
NM_102228.4 | NP_564204.1 | 3 - 4 | 425 | 110 | ||
GenBank mRNA | AY035139.1 | - | 3 - 4 | 417 | 110 | |
AY113912.1 | - | 3 - 4 | 343 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |