Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332636.1 | NP_001320353.1 | 3 - 4 | 694 | 87 | |
NM_102254.3 | NP_173818.1 | 3 - 4 | 749 | 87 | ||
GenBank mRNA | KJ676988.1 | - | 3 - 4 | 499 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |