Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCT1;3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332775.1 | NP_001321316.1 | 2 - 3 | 439 | 64 | |
NM_102528.4 | NP_174084.1 | 2 - 3 | 489 | 64 | ||
GenBank mRNA | AF344323.1 | - | 1 - 2 | 233 | 64 | |
AK117692.1 | - | 2 - 3 | 439 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Arabidopsis thaliana Probe spans exons |