Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102563.4 | NP_174119.1 | 7 - 8 | 1195 | 83 | |
GenBank mRNA | AB175035.1 | - | 7 - 8 | 889 | 83 | |
AF360166.1 | - | 7 - 8 | 948 | 83 | ||
AY113914.1 | - | 7 - 8 | 889 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |