Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102600.2 | NP_174156.1 | 7 - 8 | 1787 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |