Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334607.1 | NP_001320490.1 | 6 - 7 | 1231 | 107 | |
NM_106044.6 | NP_177525.2 | 6 - 7 | 1087 | 107 | ||
GenBank mRNA | AY070424.1 | - | 6 - 7 | 1079 | 107 | |
AY096602.1 | - | 6 - 7 | 1043 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |