Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344004.1 | NP_001330510.1 | 12 - 13 | 1328 | 138 | |
NM_001344005.1 | NP_001330511.1 | 13 - 14 | 1175 | 138 | ||
NM_001344006.1 | NP_001330508.1 | 13 - 14 | 1124 | 138 | ||
NM_001344007.1 | NP_001330509.1 | 10 - 11 | 1105 | 138 | ||
NM_122618.3 | NP_568494.1 | 13 - 14 | 1125 | 138 | ||
GenBank mRNA | AY026255.1 | - | 13 - 14 | 1022 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |