Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344004.1 | NP_001330510.1 | 2 - 3 | 393 | 115 | |
NM_001344005.1 | NP_001330511.1 | 2 - 3 | 393 | 115 | ||
NM_001344006.1 | NP_001330508.1 | 2 - 3 | 342 | 115 | ||
NM_122618.3 | NP_568494.1 | 2 - 3 | 343 | 115 | ||
GenBank mRNA | AY026255.1 | - | 2 - 3 | 240 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Arabidopsis thaliana Probe spans exons |