Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCH;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344019.1 | NP_001330469.1 | 11 - 12 | 1286 | 82 | |
NM_122644.5 | NP_198114.2 | 11 - 12 | 1302 | 82 | ||
GenBank mRNA | AB051072.1 | - | 11 - 12 | 827 | 82 | |
AK118707.1 | - | 11 - 12 | 1284 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |