Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122645.5 | NP_198115.2 | 15 - 16 | 1662 | 65 | |
GenBank mRNA | AY093156.1 | - | 15 - 16 | 1636 | 65 | |
BT008399.1 | - | 15 - 16 | 1558 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |