Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036892.1 | NP_001031969.1 | 7 - 8 | 877 | 112 | |
NM_001085170.1 | NP_001078639.1 | 7 - 8 | 866 | 112 | ||
NM_122954.4 | NP_198413.1 | 7 - 8 | 1091 | 112 | ||
GenBank mRNA | AF428319.1 | - | 7 - 8 | 871 | 112 | |
AF428461.1 | - | 7 - 8 | 871 | 112 | ||
AK317110.1 | - | 7 - 8 | 878 | 112 | ||
AY081252.1 | - | 7 - 8 | 825 | 112 | ||
AY088222.1 | - | 7 - 8 | 904 | 112 | ||
AY091114.1 | - | 7 - 8 | 873 | 112 | ||
AY122977.1 | - | 6 - 7 | 767 | 112 | ||
S69727.1 | - | 7 - 8 | 838 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |