Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123384.3 | NP_568577.1 | 2 - 3 | 1057 | 69 | |
GenBank mRNA | AY046023.1 | - | 2 - 3 | 873 | 69 | |
AY142608.1 | - | 2 - 3 | 801 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |