Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344328.1 | NP_001318713.1 | 2 - 3 | 272 | 145 | |
NM_123408.3 | NP_198860.1 | 2 - 3 | 268 | 145 | ||
GenBank mRNA | AB015314.1 | - | 2 - 3 | 134 | 145 | |
AK117136.1 | - | 2 - 3 | 218 | 145 | ||
BT006481.1 | - | 1 - 2 | 104 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |