Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123524.4 | NP_198975.1 | 1 - 2 | 396 | 83 | |
GenBank mRNA | AY035169.1 | - | 1 - 2 | 249 | 83 | |
AY057638.1 | - | 1 - 2 | 295 | 83 | ||
AY150449.1 | - | 1 - 2 | 195 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |