Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344443.1 | NP_001331624.1 | 8 - 9 | 1629 | 75 | |
NM_123604.3 | NP_199054.1 | 8 - 9 | 1632 | 75 | ||
GenBank mRNA | BT000785.1 | - | 8 - 9 | 1546 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |