Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203535.1 | NP_001190464.1 | 20 - 21 | 3349 | 128 | |
NM_001344505.1 | NP_001331600.1 | 19 - 20 | 3272 | 128 | ||
NM_001344506.1 | NP_001331599.1 | 19 - 20 | 3089 | 128 | ||
NM_123748.3 | NP_199194.1 | 19 - 20 | 3144 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |