Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125911.1 | NP_001119383.1 | 4 - 5 | 601 | 110 | |
NM_001344678.1 | NP_001332006.1 | 4 - 5 | 592 | 110 | ||
NM_124011.4 | NP_199453.2 | 4 - 5 | 595 | 110 | ||
GenBank mRNA | AY074261.1 | - | 4 - 5 | 592 | 110 | |
AY096737.1 | - | 3 - 4 | 364 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |