Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124045.5 | NP_199487.1 | 4 - 5 | 1039 | 82 | |
GenBank mRNA | AY099693.1 | - | 4 - 5 | 963 | 82 | |
AY128872.1 | - | 4 - 5 | 875 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |