Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABA5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124125.4 | NP_199563.1 | 4 - 5 | 748 | 79 | |
GenBank mRNA | AK222160.1 | - | 4 - 5 | 745 | 79 | |
AK227772.1 | - | 4 - 5 | 745 | 79 | ||
BT004809.1 | - | 1 - 2 | 340 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |