Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344745.1 | NP_001332593.1 | 7 - 8 | 647 | 84 | |
NM_124150.5 | NP_199587.1 | 7 - 8 | 647 | 84 | ||
GenBank mRNA | AK118640.1 | - | 7 - 8 | 514 | 84 | |
BT005294.1 | - | 7 - 8 | 499 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |