Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124177.4 | NP_568690.1 | 12 - 13 | 1124 | 72 | |
GenBank mRNA | AK118588.1 | - | 12 - 13 | 1031 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |