Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124335.3 | NP_199768.1 | 9 - 10 | 955 | 71 | |
GenBank mRNA | AK228156.1 | - | 9 - 10 | 875 | 71 | |
AM745095.1 | - | 9 - 10 | 811 | 71 | ||
AY140065.1 | - | 9 - 10 | 871 | 71 | ||
BT003398.1 | - | 9 - 10 | 811 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |