Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344892.1 | NP_001330206.1 | 7 - 8 | 961 | 73 | |
NM_124422.4 | NP_199851.2 | 7 - 8 | 1036 | 73 | ||
GenBank mRNA | AY842027.1 | - | 7 - 8 | 746 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |