Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXO70A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124613.4 | NP_200047.3 | 10 - 11 | 1931 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |