Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTSH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124696.4 | NP_568787.1 | 10 - 11 | 1587 | 114 | |
GenBank mRNA | AY091086.1 | - | 10 - 11 | 1515 | 114 | |
AY123027.1 | - | 10 - 11 | 1486 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |