Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124731.1 | NP_200163.1 | 2 - 3 | 588 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |