Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124731.1 | NP_200163.1 | 5 - 6 | 1605 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |