Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YSL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203603.2 | NP_001190532.1 | 3 - 4 | 821 | 151 | |
NM_001345061.1 | NP_001332081.1 | 3 - 4 | 988 | 151 | ||
NM_001345063.1 | NP_001332080.1 | 2 - 3 | 1046 | 151 | ||
NM_001345064.1 | NP_001332078.1 | 1 - 2 | 458 | 151 | ||
NM_124735.4 | NP_200167.2 | 3 - 4 | 697 | 151 | ||
GenBank mRNA | DQ385057.1 | - | 1 - 2 | 413 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |