Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124745.3 | NP_200177.1 | 2 - 3 | 1062 | 87 | |
GenBank mRNA | AB493791.1 | - | 2 - 3 | 603 | 87 | |
AK176109.1 | - | 2 - 3 | 703 | 87 | ||
BT010882.1 | - | 2 - 3 | 603 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |